EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Effects of diarylheptanoids on the tumor necrosis factor-alpha-induced expression of adhesion molecules in human umbilical vein endothelial cells.
PMID 17929893
Abstract
Atherosclerosis is a chronic inflammatory disease that is characterized by infiltration of mononuclear lymphocytes into the intima through the expression of adhesion molecules on the arterial wall. In the present study, we report the inhibitory effects of two diarylheptanoids, 5-O-methylhirsutanonol (1) and oregonin (2), isolated from the methanolic extracts of Alnus japonica leaves, on the expression of adhesion molecules in human umbilical vein endothelial cells (HUVECs). Compounds 1 and 2 inhibited tumor necrosis factor (TNF)-alpha-induced up-regulation of vascular cell adhesion molecule-1 (VCAM-1) and intercellular adhesion molecule-1 (ICAM-1), which also prevented adhesion of monocytes to HUVECs, and slightly suppressed the mRNA expression of the inflammation-associated gene interleukin-1beta (IL-1beta). A further study demonstrated the inhibitory effect of compound 1 on DNA-binding of nuclear factor kappaB (NF-kappaB) and on the phosphorylation and degradation of inhibitory factor kappaBalpha (IkappaBalpha) in TNF-alpha-stimulated HUVECs. These results indicate that compounds 1 and 2 may be useful in the prevention and treatment of atherosclerosis through attenuation of adhesion molecule expression by inhibition of NF-kappaB activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00088
Oregonin, ≥95% (LC/MS-ELSD)
C24H30O10