EMAIL THIS PAGE TO A FRIEND
FEBS letters
Hydrolysis of non-cognate aminoacyl-adenylates by a class II aminoacyl-tRNA synthetase lacking an editing domain.
PMID 17931630
Abstract
Aminoacyl-tRNA synthetases, a group of enzymes catalyzing aminoacyl-tRNA formation, may possess inherent editing activity to clear mistakes arising through the selection of non-cognate amino acid. It is generally assumed that both editing substrates, non-cognate aminoacyl-adenylate and misacylated tRNA, are hydrolyzed at the same editing domain, distant from the active site. Here, we present the first example of an aminoacyl-tRNA synthetase (seryl-tRNA synthetase) that naturally lacks an editing domain, but possesses a hydrolytic activity toward non-cognate aminoacyl-adenylates. Our data reveal that tRNA-independent pre-transfer editing may proceed within the enzyme active site without shuttling the non-cognate aminoacyl-adenylate intermediate to the remote editing site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4503
DL-Serine hydroxamate, seryl-tRNA synthetase inhibitor
C3H8N2O3