EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Phenylpiperidine-benzoxazinones as urotensin-II receptor antagonists: synthesis, SAR, and in vivo assessment.
PMID 17933531
Abstract
Various 4-phenylpiperidine-benzoxazin-3-ones were synthesized and biologically evaluated as urotensin-II (U-II) receptor antagonists. Compound 12i was identified from in vitro evaluation as a low nanomolar antagonist against both rat and human U-II receptors. This compound showed in vivo efficacy in reversing the ear-flush response induced by U-II in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
639869
4-Phenylpiperidine, 97%
C11H15N