EMAIL THIS PAGE TO A FRIEND
Skin therapy letter
New developments in hormonal therapy for acne.
PMID 17940710
Abstract
Oral contraceptives (OCs) are a valuable option for the treatment of women with acne. The use of OCs can be considered across the spectrum of acne disease severity in women. In Canada, three preparations are approved for mild-to-moderate acne, and a fourth is indicated for severe acne. These formulations contain estrogen in the form of ethinyl estradiol and a progestin. In Canada, the most recently approved OC is ethinyl estradiol 0.03 mg and drospirenone 3mg (Yasmin, Bayer). With the accumulating evidence on the efficacy and safety of drospirenone-containing hormonal preparations, this formulation provides dermatologists with a new treatment option for acne and other hyperandrogenic disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94497
Norgestimate, analytical standard
C23H31NO3
SML0359
Norgestimate, ≥97% (HPLC)
C23H31NO3