EMAIL THIS PAGE TO A FRIEND
Biochimie
Production of D-phenylglycine-related amino acids by immobilized microbial cells.
PMID 17941449
Abstract
Bacterial dihydropyrimidinase was shown to catalyze the hydrolytic cleavage of various 5-substituted hydantoins to the corresponding N-carbamyl-D-amino acids under alkaline conditions. Therefore, an enzymatic method for preparing the D-forms of phenylglycine-related amino acids was developed using immobilized bacterial cells with high enzyme activity. Alkalophilic bacteria were a good enzyme source for this process. The process is simple and economical for use in the production of various amino acids with the D-configuration.