EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A new, stereoselective, ring-forming reaction of 1,2-ethanedithiol with N-acylated indoles.
PMID 17941672
Abstract
While attempting to prepare 2,5-dithiacyclopentyl derivatives from N-acyl 5-fluoroindole by reaction with 1,2-ethanedithiol we discovered that, instead of the expected product, annelation occurred to give a tricyclic compound containing a 3,6-dithiaazepine ring. This reaction is stereoselective and was found to be general for N-acylindoles, not being affected by substituents on the indole ring.
Related Materials