EMAIL THIS PAGE TO A FRIEND
The Malaysian journal of pathology
Detection of Clostridium difficile from an enhancement broth by gas-liquid chromatography.
PMID 1795558
Abstract
Gas-Liquid chromatography (GLC) was used to detect the presence of isocaproic acid produced by Clostridium difficile from 54 stool samples grown in cycloserine-cefoxitin broth. Isocaproic acid was detected in 12 samples of which 5 were confirmed to be Clostridium difficile by culture and biochemical tests. The detection of isocaproic acid by GLC together with the presence of presumptive colonies on primary selective culture media provides a more rapid laboratory diagnosis for Clostridium difficile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W346306
4-Methylpentanoic acid, ≥98%, FCC, FG
C6H12O2
277827
4-Methylvaleric acid, 99%
C6H12O2