EMAIL THIS PAGE TO A FRIEND
Nucleic Acids Research
An RNA targeted to the HIV-1 LTR promoter modulates indiscriminate off-target gene activation.
PMID 17959645
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody