EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of some new anils: part 1. Reaction of 2-hydroxy-benzaldehyde and 2-hydroxynaphthaldehyde with 2-aminopyridene and 2-aminopyrazine.
PMID 17960088
Abstract
New Schiff bases derived from 2-aminopyridene and 2-aminopyrazine have been synthesized. The UV-Visible spectra of the compounds have been investigated in acetonitrile and toluene. The compounds were in tautomeric equilibrium (enol-imine O- H...N, keto-amine O...H-N forms) in polar and nonpolar solvents. For some derivatives the keto-amine form was observed in both toluene and acetonitrile. 1H-NMR and IR results showed that all Schiff bases studied favor the enol-imine form over the keto form in a weakly polar solvent such as deuterochloroform.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A76958
Aminopyrazine, 98%
C4H5N3