EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of new trans-2-benzyl-3-(furan-2-yl)-4-substituted-1,2,3,4-tetrahydroisoquinolinones.
PMID 17962773
Abstract
The reaction of homophthalic anhydride and N-(furan-2-yl-methylidene)- benzylamine in different solvents and varying temperatures was studied in detail. Mixtures of the expected trans- and cis-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acids trans-5 and cis-5, along with by-products 6 and 7 were obtained in dichloroethane or benzene. In pyridine, used for the first time, the reaction became completely diastereoselective, giving only the trans isomer. The carboxylic acid group of trans-5 was transformed in four steps into cyclic aminomethyl groups which yielded various new tetrahydroisoquinolinones trans- 11a-g, incorporating both a known fragment of pharmacological interest and various pharmacophoric substituents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128589
Homophthalic anhydride, technical grade
C9H6O3