EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Liquid chromatography-atmospheric pressure photoionization-mass spectrometry analysis of triacylglycerol lipids--effects of mobile phases on sensitivity.
PMID 17964589