EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Expression of a Gi-coupled receptor in the heart causes impaired Ca2+ handling, myofilament injury, and dilated cardiomyopathy.
PMID 17965283
Abstract
Increased signaling by G(i)-coupled receptors has been implicated in dilated cardiomyopathy. To investigate the mechanisms, we used transgenic mice that develop dilated cardiomyopathy after conditional expression of a cardiac-targeted G(i)-coupled receptor (Ro1). Activation of G(i) signaling by the Ro1 agonist spiradoline caused decreased cellular cAMP levels and bradycardia in Langendorff-perfused hearts. However, acute termination of Ro1 signaling with the antagonist nor-binaltorphimine did not reverse the Ro1-induced contractile dysfunction, indicating that Ro1 cardiomyopathy was not due to acute effects of receptor signaling. Early after initiation of Ro1 expression, there was a 40% reduction in the abundance of the sarcoplasmic reticulum Ca(2+)-ATPase (P < 0.05); thereafter, there was progressive impairment of both Ca(2+) handling and force development assessed with ventricular trabeculae. Six weeks after initiation of Ro1 expression, systolic Ca(2+) concentration was reduced to 0.61 +/- 0.08 vs. 0.91 +/- 0.07 microM for control (n = 6-8; P < 0.05), diastolic Ca(2+) concentration was elevated to 0.41 +/- 0.07 vs. 0.23 +/- 0.06 microM for control (n = 6-8; P < 0.01), and the decline phase of the Ca(2+) transient (time from peak to 50% decline) was slowed to 0.25 +/- 0.02 s vs. 0.13 +/- 0.02 s for control (n = 6-8; P < 0.01). Early after initiation of Ro1 expression, there was a ninefold elevation of matrix metalloproteinase-2 (P < 0.01), which is known to cause myofilament injury. Consistent with this, 6 wk after initiation of Ro1 expression, Ca(2+)-saturated myofilament force in skinned trabeculae was reduced to 21 +/- 2 vs. 38 +/- 0.1 mN/mm(2) for controls (n = 3; P < 0.01). Furthermore, electron micrographs revealed extensive myofilament damage. These findings may have implications for some forms of human heart failure in which increased activity of G(i)-coupled receptors leads to impaired Ca(2+) handling and myofilament injury, contributing to impaired ventricular pump function and heart failure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U105
U-62066, solid
C22H30Cl2N2O2 · CH4O3S