EMAIL THIS PAGE TO A FRIEND
Acta biochimica Polonica
Characterization of acyllipid: sterol glucoside acyltransferase from oat (Avena sativa L.) seedlings.
PMID 1796705
Abstract
Membranous fractions from leaves of oat seedlings readily convert cholesterol beta-D-glucoside into its 6'-O-acyl derivative using endogenous acyllipids as acyl sources. Experiments with delipidated enzyme preparations showed that among acyllipids present in oat leaves digalactosyldiacylglycerols are evidently the best acyl donors in this reaction. Beside of sterol glucosides, the enzyme can acylate beta-D-glucosides of several other steroids, although at very different rates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28609
Cholesterol β-D-glucoside, ≥97% (TLC)
C33H56O6