EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
One-pot cyclization of 2-aminophenethyl alcohols: a novel and direct approach to the synthesis of N-acyl indolines.
PMID 17973530
Abstract
A unique one-pot cyclization of 2-aminophenethyl alcohols with carboxylic acids in the presence of PPh3, CCl4, and NEt3 furnished the formation of N-acyl indolines in good to excellent yields. This new approach provides an efficient, scalable, low-cost, and direct access to the biologically important indolines which are further oxidizable to indoles and oxindoles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
192600
2-Aminophenethyl alcohol, 97%
C8H11NO