EMAIL THIS PAGE TO A FRIEND
Medicinal research reviews
Recent advances in plant hepatoprotectives: a chemical and biological profile of some important leads.
PMID 17979145
Abstract
Medicinal plants have been traditionally used for treating liver diseases since centuries. Several leads from plant sources have been found as potential hepatoprotective agents with diverse chemical structures. Although, a big list of hepatoprotective phytomolecules was reported in the scientific literature, only a few were potent against various types of liver damages. Of which, silymarin, andrographolide, neoandrographolide, curcumin, picroside, kutkoside, phyllanthin, hypophyllanthin, and glycyrrhizin have largely attracted the scientific community. This review focuses discussion on the chemistry, biological activity, mode of action, toxicity, and future prospects of these leads.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49879
Neoandrographolide, analytical standard
C26H40O8