EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Biphenyl amide p38 kinase inhibitors 2: Optimisation and SAR.
PMID 17981461
Abstract
The biphenyl amides are a novel series of p38 MAP kinase inhibitors. Structure-activity relationships of the series against p38alpha are discussed with reference to the X-ray crystal structure of an example. The series was optimised rapidly to a compound showing oral activity in an in vivo disease model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
264660
3-Bromo-4-methylbenzoic acid, technical grade, 85%
C8H7BrO2