EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
AMPA and kainate receptors mediate mutually exclusive effects on GABA(A) receptor expression in cultured mouse cerebellar granule neurones.
PMID 17986225
Abstract
Studies on animal models of epilepsy and cerebellar ataxia, e.g., stargazer mice (stg) have identified changes in the GABAergic properties of neurones associated with the affected brain loci. Whether these changes contribute to or constitute homeostatic adaptations to a state of altered neuronal excitability is as yet unknown. Using cultured cerebellar granule neurones from control [+/+; alpha-amino-3-hydroxyl-5-methyl-4-isoxazolepropionate receptor (AMPAR)-competent, Kainate receptor (KAR)-competent] and stg (AMPAR-incompetent, KAR-competent), we investigated whether non-NMDA receptor (NMDAR) activity regulates GABA(A) receptor (GABAR) expression. Neurones were maintained in 5 mmol/L KCl-containing basal media or depolarizing media containing either 25 mmol/L KCl or the non-NMDAR agonist kainic acid (KA) (100 micromol/L). KCl- and KA-mediated depolarization down-regulated GABAR alpha1, alpha6 and beta2, but up-regulated alpha4, beta3 and delta subunits in +/+ neurones. The KCl-evoked but not KA-evoked effects were reciprocated in stg neurones compatible with AMPAR-regulation of GABAR expression. Conversely, GABAR gamma2 expression was insensitive to KCl-mediated depolarization, but was down-regulated by KA-treatment in a 6-cyano-7-nitroquinoxaline-2,3-dione (CNQX)-reversible manner in +/+ and stg neurones compatible with a KAR-mediated response. KA-mediated up-regulation of GABAR alpha4, beta3 and delta was inhibited by L-type voltage-gated calcium channel (L-VGCC) blockers and the Ca2+/calmodulin-dependent protein kinase inhibitor, 4-[(2S)-2-[(5-isoquinolinylsulfonyl)methylamino]-3-oxo-3-(4-phenyl-1-piperazinyl)propyl] phenyl isoquinoline sulfonic acid ester (KN-62). Up-regulation of GABAR alpha4 and beta3 was also prevented by calcineurin (CaN) inhibitors, FK506 and cyclosporin A. Down-regulation of GABAR alpha1, alpha6 and beta2 was independent of L-VGCC activity, but was prevented by inhibitors of CaN. Thus, we provide evidence that a KAR-mediated and at least three mutually exclusive AMPAR-mediated signalling mechanisms regulate neuronal GABAR expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R109
Ro 15-4513, solid
C15H14N6O3