EMAIL THIS PAGE TO A FRIEND
Patologicheskaia fiziologiia i eksperimental'naia terapiia
[The prevention of the development of the collagen-induced microembolic syndrome by decreasing the hydrodynamic resistance of the blood with special polymers under flow].
PMID 1798660
Abstract
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced microembolic syndrome and the effect produced on it by hydrodynamic resistance reducing polymers under conditions modelling real circulation. The authors show the formation of a developed microembolic syndrome under the effect of collagen with accumulation of vasoconstrictors in the environment and increase of red cell hemolysis. Reduction of blood flow turbulence by the addition of solutions of neutral high-molecular linear polymers revealed the inhibiting effect of these polymers on the development of intracellular aggregate formation and concomitant red cell microhemolysis, which was attended by sharp reduction of thromboxane A2 production in the experiment. The authors show the significance of hemodynamic conditions in the development of the microembolic syndrome and the possibility of preventing it under conditions of modification of the blood flow with polymers, which may be of high prophylactic importance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91462
Kollisolv® PEG E 300
06855
Kollisolv® PEG E 400
89510
Poly(ethylene glycol), BioUltra, 8,000
202398
Poly(ethylene glycol), average Mn 400
P5413
Poly(ethylene glycol), for molecular biology, average mol wt 8,000
81260
Poly(ethylene glycol), average Mn 6,000
P3015
Poly(ethylene glycol), average mol wt 200
P2139
Poly(ethylene glycol), average mol wt 8,000, powder
P4338
Poly(ethylene glycol), BioXtra, average mol wt 3,350, powder
202444
Poly(ethylene glycol), average Mn 3,350, powder
95904
Poly(ethylene glycol), BioUltra, 4,000
81268
Poly(ethylene glycol), BioUltra, for molecular biology, 8,000
81300
Poly(ethylene glycol), average Mn 20,000
81255
Poly(ethylene glycol), BioUltra, 6,000
95172
Poly(ethylene glycol), BioUltra, 20,000
81240
Poly(ethylene glycol), average Mn 4,000, platelets
81310
Poly(ethylene glycol), 35,000
81253
Poly(ethylene glycol), BioUltra, for molecular biology, 6,000
81172
Poly(ethylene glycol), tested according to Ph Eur, 400
202371
Poly(ethylene glycol), average Mn 300
P3265
Poly(ethylene glycol), average mol wt 400
91893
Poly(ethylene glycol), BioUltra, 400
84797
Poly(ethylene glycol), BioUltra, 2,000
88276
Poly(ethylene glycol), BioUltra, 3,350
94646
Poly(ethylene glycol), BioUltra, 35,000
81280
Poly(ethylene glycol), 10,000
81188
Poly(ethylene glycol), BioUltra, 1,000
92897
Poly(ethylene glycol), BioUltra, 10,000
202401
Poly(ethylene glycol), average Mn 600, waxy solid (moist)
90878
Poly(ethylene glycol), BioUltra, 300