EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Longer guests drive the reversible assembly of hyperextended capsules.
PMID 17987592
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7305
Glycoluril, 97%
C4H6N4O2