EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
First total syntheses and spectral data corrections of 11-alpha-methoxycurvularin and 11-beta-methoxycurvularin.
PMID 17988150
Abstract
Concise and efficient total syntheses of 11-alpha-methoxycurvularin and 11-beta-methoxycurvularin were accomplished for the first time. The three-component linchpin coupling and intramolecular acylation reactions were key steps, in which we found the spectral data of 11-alpha-methoxycurvularin and 11-beta-methoxycurvularin, reported in the literature, were reversed with each other.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0772
Curvularin, from Penicillium citrinum, ≥98% (HPLC)
C16H20O5