EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Oxygen-17 hyperfine structures in the pure rotational spectra of SrO, SnO, BaO, HfO and ThO.
PMID 17989798
Abstract
Hyperfine structures arising from the couplings of the nuclear spin angular momentum of (17)O (I = 5/2) with the end over end rotation of several metal-containing diatomic monoxides have been observed using a Fourier transform microwave spectrometer. The molecules have been produced by reacting (17)O(2) with laser ablated metal atoms. The oxygen-17 nuclear quadrupole coupling constants have been determined for the title molecules and are interpreted in terms of a simple Townes-Dailey model. Also, the oxygen-17 nuclear spin-rotation constants have been determined and used to calculate the oxygen-17 shieldings for each molecule.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
554847
Barium oxide, 99.99% trace metals basis
BaO
288497
Barium oxide, 97%
BaO