EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
DNA methylation inhibitor 5-Aza-2'-deoxycytidine induces reversible genome-wide DNA damage that is distinctly influenced by DNA methyltransferases 1 and 3B.
PMID 17991895