EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Cooperation of translocase complexes in mitochondrial protein import.
PMID 17998403
Abstract
Most mitochondrial proteins are synthesized in the cytosol and imported into one of the four mitochondrial compartments: outer membrane, intermembrane space, inner membrane, and matrix. Each compartment contains protein complexes that interact with precursor proteins and promote their transport. These translocase complexes do not act as independent units but cooperate with each other and further membrane complexes in a dynamic manner. We propose that a regulated coupling of translocases is important for the coordination of preprotein translocation and efficient sorting to intramitochondrial compartments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500987
Anti-TOM70 antibody produced in rabbit, affinity isolated antibody