EMAIL THIS PAGE TO A FRIEND
Acta paediatrica (Oslo, Norway : 1992)
Pulmonary function tests in an infant with osteogenesis imperfecta and early biphosphonate treatment.
PMID 18001342
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N6037
Neridronate, ≥97% (NMR), solid
C6H17NO7P2