EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Magnetic nanoparticle-supported proline as a recyclable and recoverable ligand for the CuI catalyzed arylation of nitrogen nucleophiles.
PMID 18004449
Abstract
Magnetic nanoparticle-supported proline ligand was prepared and used for the CuI catalyzed Ullmann-type coupling reactions of aryl/heteroaryl bromides with various nitrogen heterocycles to form the corresponding N-aryl products in good to excellent yields; furthermore, this magnetic nanoparticle-supported proline ligand could be readily separated using an external magnet and reused without significant loss of activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B56404
4′-Bromoacetophenone, 98%
C8H7BrO