EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Reactivity of Pd(II) complexes with electrophilic chlorinating reagents: isolation of Pd(IV) products and observation of C-Cl bond-forming reductive elimination.
PMID 18004863