EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Microbial hydroxylation of sclareol by Rhizopus stolonifer.
PMID 18007369
Abstract
Incubation of sclareol with Rhizopus stolonifer affords in high yield a mixture of triols with 18-hydroxy-sclareol as the main component.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357995
Sclareol, 98%
C20H36O2