EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Improved synthesis of beta-D-6-methylpurine riboside and antitumor effects of the beta-D- and alpha-D-anomers.
PMID 18007371
Abstract
6-Methylpurine-beta-D-riboside (beta-D-MPR) has been synthesized by coupling 6-methylpurine and 1-O-acetyl-2,3,5-tri-O-benzoyl-D-ribose using conditions that produce the beta-D-anomer exclusively. The in vitro antitumor effects of beta-D-MPR and 6-methyl-purine-alpha-D-riboside (alpha-D-MPR) in five human tumor cell lines showed that beta-D-MPR was highly active (IC(50) values ranging from 6 to 34 nM). alpha-D-MPR, although less active than beta-D-MPR, also exhibited significant antitumor effects (IC50 values ranging from 1.47 to 4.83 microM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4002
6-Methylmercaptopurine riboside, ≥99% (HPLC)
C11H14N4O4S