EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and spectroscopic characterization of new Schiff bases containing the benzo-15-crown-5 moiety.
PMID 18007486
Abstract
New crown ether Schiff base derivatives were prepared by the condensation of 4'-formylbenzo-15-crown-5 or 4'-formyl-5'-hydroxybenzo-15-crown-5 with 1,2-bis(2-aminophenoxy)ethane. The structures of these new compounds were confirmed on the basis of elemental analysis, IR, (1)H- and (13)C-NMR, UV-Vis and mass spectroscopic data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
188832
15-Crown-5, 98%
C10H20O5