EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and biological activities of novel triazole compounds containing 1,3-dioxolane rings.
PMID 18007492
Abstract
Thirteen new triazoles containing 1,3-dioxolane rings were synthesized and their identities confirmed by means of IR, NMR, MS, elemental analysis and X-ray crystallography. The results of preliminary biological tests show that all of these compounds possess some fungicidal and plant growth regulant activities.