EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of novel, potentially biologically active dibenzosuberone derivatives.
PMID 18007539
Abstract
Novel representatives of the important group of biologically active dibenzosuberone derivatives were prepared: 3,7-dibromo-5-(dimethylaminoethyl- oxyimino)-10,11-dihydro-5H-dibenzo[a,d]cyclohepta-1,4-diene (1), 3,7-dibromo-5-(3- dimethylaminopropylidene)-10,11-dihydro-5H-dibenzo[a,d]cycloheptene (2) and 1,7- dibromo-5-(3-dimethylaminopropylidene)-10,11-dihydro-5H-dibenzo-[a,d]-cycloheptene (3). These compounds are potential tricyclic antidepressants (TCAs), which are still the most frequently prescribed antidepressants in many countries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1000000
Dibenzosuberone, European Pharmacopoeia (EP) Reference Standard
C15H12O