EMAIL THIS PAGE TO A FRIEND
CNS spectrums
A seven- year follow-up of an extremely refractory bipolar I patient.
PMID 18018326
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F112
(+)-Fenfluramine hydrochloride
C12H16F3N · HCl