EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
One-solvent extraction of astaxanthin from lactic acid fermented shrimp wastes.
PMID 18020413
Abstract
Free astaxanthin one-solvent extractions with ethanol, acetone, and liquid 1,1,1,2-tetrafluoroethane from raw and lactic acid fermented (ensilaged) shrimp residues were investigated. The total carotenoid recovery from ensilaged shrimp wastes was higher than that from non-ensilaged ones as assessed by HPLC analyses. Acetone gave the highest extraction yields of free astaxanthin with up to 115 microg/g of material. Moreover, liquid tetrafluoroethane is reported for the first time in a successful one-solvent extraction of carotenoids from shrimp.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
374334
1,1,1,2-Tetrafluoroethane, ≥99%
C2H2F4