EMAIL THIS PAGE TO A FRIEND
FEBS letters
The lipin protein family: dual roles in lipid biosynthesis and gene expression.
PMID 18023282
Abstract
The prevalence of obesity in the western world has focused attention on factors that influence triglyceride biosynthesis, storage, and utilization. Members of the lipin protein family have a newly discovered enzymatic role in triglyceride and phospholipid biosynthesis as a phosphatidate phosphatase, and also act as an inducible transcriptional coactivator in conjunction with peroxisome proliferator-activated receptor gamma (PPAR gamma) coactivator-1 alpha and PPAR alpha. Through these activities, the founding member of the family, lipin-1, influences lipid metabolism and glucose homeostasis in diverse tissues including adipose tissue, skeletal muscle, and liver. The physiological roles of lipin-2 and lipin-3 are less well defined, but are likely to carry out similar functions in glycerolipid biosynthesis and gene expression in a distinct tissue distribution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3501040
Anti-LPIN1 antibody produced in rabbit, affinity isolated antibody