EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Neuroprotective effects of ginseng saponins against L-type Ca2+ channel-mediated cell death in rat cortical neurons.
PMID 18023426
Abstract
In the present study, we have examined any possible involvement of L-type Ca(2+) channels in ginseng-mediated neuroprotective actions. Exposure to a 50mM KCl (high-K) produced neuronal cell death, which was blocked by a selective L-type Ca(2+) channel blocker in cultured cortical neurons. When cultured cells were co-treated with ginseng total saponin (GTS) and high-K, GTS reduced high-K-induced neuronal death. Using Ca(2+) imaging techniques, we found that GTS inhibited high-K-mediated acute and long-term [Ca(2+)](i) changes. These GTS-mediated [Ca(2+)](i) changes were diminished by nifedipine. Furthermore, GTS-mediated effects were also diminished by a saturating concentration of Bay K (10muM). After confirming the protective effect of GTS using a TUNEL assay, we found that ginsenosides Rf and Rg(3) are active components in ginseng-mediated neuroprotection. These results suggest that inhibition of L-type Ca(2+) channels by ginseng could be one of the mechanisms for ginseng-mediated neuroprotection in cultured rat cortical neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
65839
Ginsenoside Rf, analytical standard
C42H72O14
01580590
Ginsenoside Rf, primary reference standard
C42H72O14