EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
Suppression of the HPA axis in pediatric patients with atopic dermatitis.
PMID 18025377
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0099
Fluocinonide, ≥98% (HPLC)
C26H32F2O7