EMAIL THIS PAGE TO A FRIEND
Organic letters
New synthetic path to 2,2'-bipyridine-5,5'-dicarbaldehyde and its use in the [3+3] cyclocondensation with trans-1,2-diaminocyclohexane.
PMID 18027964
Abstract
2,2'-Bipyridine-5,5'-dicarbaldehyde has been prepared in two steps by enamination of 5,5'-dimethyl-2,2'-bipyridine with Bredereck's reagent, and subsequent oxidative cleavage of the enamine groups with sodium periodate. On condensation of this dialdehyde with enantiomerically pure trans-1,2-diaminocyclohexane, the macrocyclic [3+3] hexa Schiff base has been obtained in excellent yield. Its reduction has given large macrocyclic hexaamine having three bipyridine units incorporated into the macrocycle structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270016
(±)-trans-1,2-Diaminocyclohexane, 99%
C6H14N2