EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series (2004)
Synthesis of DNA duplexes containing complexes of 5-modified pyrimidine bases and metal ions.
PMID 18029646
Abstract
DNA duplexes carrying metal ions at the 5-position of uracil residues were synthesized by mixing oligodeoxyribonucleotides (ODNs) containing 5-formyl uracils, metal ions, and amines. A metal ion binding site may form from the 5-formyl residue and amines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414174
5-Formyluracil, 98%
C5H4N2O3