EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series (2004)
Suppression of subgenomic hepatitis C virus replication by 5'-O-masked analogues of 6-chloropurine-2'-deoxyriboside.
PMID 18029775
Abstract
A series of nucleoside analogues whose 5'-hydroxyl groups are masked by various protective groups were synthesized and evaluated to develop novel anti- hepatitis C virus (HCV) agents. Among the several analogues that showed anti-HCV potency, a 5'-O-benzoyl-2'-deoxyribonucleoside analogue exhibited high anti-HCV activity with an EC(50) of 6.1 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161179
6-Chloropurine, ≥99%
C5H3ClN4
511617
6-Chloropurine, 99%
C5H3ClN4
26260
6-Chloropurine, ≥95.0% (HPLC)
C5H3ClN4