EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on isoamyl salicylate.
PMID 18031914
Abstract
A toxicologic and dermatologic review of isoamyl salicylate when used as a fragrance ingredient is presented.