EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Optochin resistance among Streptococcus pneumoniae strains colonizing healthy children in Portugal.
PMID 18032618
Abstract
Two percent of 1,973 pneumococcus strains isolated from carriers since 2001 in Portugal were found to be optochin resistant. These strains belonged to eight serotypes (and some were nontypeable), and they had diverse genetic backgrounds. Novel optochin-resistant lineages were detected over time, suggesting that there was a continuous, although sporadic, emergence of optochin resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8253
Ethylhydrocupreine, analytical standard
C21H28N2O2
E9876
Ethylhydrocupreine hydrochloride, analytical standard
C21H28N2O2 · HCl