EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Biochemistry. Signaling across the cell membrane.
PMID 18033872
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53787
Carazolol, analytical standard
C18H22N2O2