EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on cinnamyl isobutyrate.
PMID 18035465
Abstract
A toxicologic and dermatologic review of cinnamyl isobutyrate when used as a fragrance ingredient is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W229709
Cinnamyl isobutyrate, ≥97%, FCC, FG
C13H16O2