EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on benzyl salicylate.
PMID 18035470
Abstract
A toxicologic and dermatologic review of benzyl salicylate when used as a fragrance ingredient is presented.