EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on alpha-amylcinnamyl alcohol.
PMID 18037205
Abstract
A toxicologic and dermatologic review of alpha-amylcinnamyl alcohol when used as a fragrance ingredient is presented.