EMAIL THIS PAGE TO A FRIEND
Chemosphere
Levels and spatial distribution of chlorophenols - 2,4-dichlorophenol, 2,4,6-trichlorophenol, and pentachlorophenol in surface water of China.
PMID 18037470
Abstract
The chlorophenol pollutants (CPs) have been reported to occur at relatively high concentrations in some Chinese waters. To map the distribution of CPs in the surface water throughout China, samples were collected from over 600 sites in the seven major watersheds and three drainage areas. The samples were analyzed for the representative CPs including 2,4-dichlorophenol, 2,4,6-trichlorophenol, and pentachlorophenol. In general, it was observed that 2,4-dichlorophenol and 2,4,6-trichlorophenol were more frequently detected at higher concentrations in the rivers of North China compared with those of South China. High concentration sites of 2,4-dichlorophenol and 2,4,6-trichlorophenol mainly occurred in the Yellow River, Huaihe River, and Haihe River watersheds, while pentachlorophenol contamination mainly occurred in the Yangtze River watershed. The pentachlorophenol was the most ubiquitous CPs being detected in 85.4% of samples (median=50.0ngl(-1); range <1.1-594.0ngl(-1)), 2,4-dichlorophenol was detected in 51.3% (median=5.0ngl(-1); range <1.1-19960.0ngl(-1)) and the 2,4,6-trichlorophenol was detected in 54.4% of water samples (median=2.0ngl(-1), range <1.4-28650.0ngl(-1)). The results of this investigation indicated that 2,4-dichlorophenol and 2,4,6-trichlorophenol contaminations of Yellow River, Huaihe River, and Haihe River watersheds were of particular concern, while the pentachlorophenol contamination mainly occurred in the Yangtze River watershed. These results showed that CPs contamination in the surface water of China was similar to other places of the world.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40870
1,2-Dimethyl-3-nitrobenzene, Selectophore, ≥99.0%
C8H9NO2