EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Copper-catalyzed [4 + 1] cycloadditions of alpha,beta-acetylenic ketones with diazoacetates to form trisubstituted furans.
PMID 18041850
Abstract
Copper-catalyzed [4 + 1] cycloaddition reaction of alpha,beta-acetylenic ketones with alpha-diazo esters offers an efficient, direct route to highly substituted furans. The reaction conditions and the scope of the process are examined, and a possible mechanism is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2