EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Leucobacter chromiireducens subsp. solipictus subsp. nov., a pigmented bacterium isolated from the nematode Caenorhabditis elegans, and emended description of L. chromiireducens.
PMID 18048723
Abstract
A yellow-pigmented, Gram-positive, aerobic, non-motile, non-spore-forming, irregular rod-shaped bacterium (strain TAN 31504(T)) was isolated from the bacteriophagous nematode Caenorhabditis elegans. Based on 16S rRNA gene sequence similarity, DNA G+C content of 69.5 mol%, 2,4-diaminobutyric acid in the cell-wall peptidoglycan, major menaquinone MK-11, abundance of anteiso- and iso-fatty acids, polar lipids diphosphatidylglycerol and phosphatidylglycerol and a number of shared biochemical characteristics, strain TAN 31504(T) was placed in the genus Leucobacter. DNA-DNA hybridization comparisons demonstrated a 91 % DNA-DNA relatedness between strain TAN 31504(T) and Leucobacter chromiireducens LMG 22506(T) indicating that these two strains belong to the same species, when the recommended threshold value of 70 % DNA-DNA relatedness for the definition of a bacterial species by the ad hoc committee on reconciliation of approaches to bacterial systematics is considered. Based on distinct differences in morphology, physiology, chemotaxonomic markers and various biochemical characteristics, it is proposed to split the species L. chromiireducens into two novel subspecies, Leucobacter chromiireducens subsp. chromiireducens subsp. nov. (type strain L-1(T)=CIP 108389(T)=LMG 22506(T)) and Leucobacter chromiireducens subsp. solipictus subsp. nov. (type strain TAN 31504(T)=DSM 18340(T)=ATCC BAA-1336(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32830
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
C4H10N2O2 · 2HCl