EMAIL THIS PAGE TO A FRIEND
The American journal of nursing
How to try this: detecting delirium.
PMID 18049062
Abstract
For patients and their loved ones, delirium can be a frightening experience. A fluctuating mental status is important to identify because it often signals a need for additional treatment. The Confusion Assessment Method (CAM) diagnostic algorithm enables nurses to assess for delirium by identifying the four features of the disorder that distinguish it from other forms of cognitive impairment. It can be completed in five minutes and is easily incorporated into ongoing assessments of hospitalized patients. (This screening tool is included in the series Try This: Best Practices in Nursing Care to Older Adults, from the Hartford Institute for Geriatric Nursing at New York University's College of Nursing.) For a free online video demonstrating the use of this tool, go to http://links.lww.com/A209.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7012
Guaiacol glyceryl ether carbamate, analytical standard
C11H15NO5