EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Telmisartan improves endothelial function in patients with essential hypertension.
PMID 18049303
Abstract
: Hypertension is a cardiovascular risk factor commonly associated with endothelial dysfunction and increased renal vascular resistance. Angiotensin receptor blockers (ARBs) may beneficially affect these parameters via antagonism of angiotensin type 1 (AT1) receptor-mediated vasoconstriction and vascular superoxide production. We therefore investigated whether the new ARB telmisartan improves endothelial function and renal vascular resistance in patients with essential hypertension. : Thirty-seven patients with essential hypertension were randomized to receive telmisartan, the calcium channel blocker nisoldipine, or their combination for 6 weeks in a prospective, parallel group study. Brachial artery flow-mediated (endothelium-dependent) dilation (FMD) and renal vascular resistance index (RVRI) were evaluated using high-resolution ultrasound before, at 3 weeks (low dose), and at 6 weeks (high dose) after initiation of treatment. : At baseline, FMD and RVRI did not significantly differ between treatment groups. After 3 weeks of treatment neither treatment significantly changed FMD or RVRI. After 6 weeks of treatment, patients randomized to receive telmisartan alone or the combination, but not those treated with nisoldipine alone, displayed a significantly improved FMD, whereas RVRI values again were not significantly different as compared to those at baseline. : In our study cohort of patients with essential hypertension, treatment with telmisartan improved FMD but did not change RVRI. Future studies will demonstrate whether this telmisartan-induced effect may contribute to a blood pressure-independent reduction in cardiovascular morbidity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0165
Nisoldipine, ≥98% (HPLC)
C20H24N2O6